missing translation for 'onlineSavingsMsg'
Learn More

Spearmint Oil, Natural, FCC, 55%, Spectrum™ Chemical

SP306, 8008-79-5

Supplier:  Spectrum Chemical Mfg Cor SP30625LBBL

 View more versions of this product

Catalog No. 18606971


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Spearmint Oil, Natural, FCC is used for cooking, flavoring, and as a digestive support. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

8008-79-5
Steel Pail
25 lb.
FCC
1
1.484-1.491
55%

Chemical Identifiers

8008-79-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions