missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ Specialty Glass Column: Glass Coarse Frit

Described in US EPA Protocol - Fill columns with appropriate adsorbents when cleaning or drying environmental samples for pesticide or priority pollutant analyses

Supplier:  MilliporeSigma™ Supelco™ 64758U

 View more versions of this product

Catalog No. 111010324


Explore available promotions

Description

Description

Specifications

Specifications

Specialty Glass Column (SJ Inlet Joint)
LPLC
1
22 mm
25 mm
Glass (Frit), PTFE (Stopcock)
Overall Length: 415 mm
Frit
Videos
SDS
Documents

Documents

Promotions

Promotions