missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spectinomycin Sulfate, ≥98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215899305

 View more versions of this product

Catalog No. ICN15899305


Explore available promotions

Description

Description

Specifications

Specifications

Spectinomycin Sulfate
Powder
White
≥98%
MFCD00270185
XGBFWQUQYQIFLB-MTTMTQIXSA-N
(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
64771
430.4
Spectinomycin Sulfate
23312-56-3
5 g
C14H26N2O11S
U 18409E, antibiotic
OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
430.43
CHEBI:9216

Chemical Identifiers

23312-56-3
430.43
XGBFWQUQYQIFLB-MTTMTQIXSA-N
CHEBI:9216
OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
C14H26N2O11S
MFCD00270185
U 18409E, antibiotic
(1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
Videos
SDS
Documents

Documents

Promotions

Promotions