missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Spectrum™ Spectra/Por™ RC Dialysis Discs
Supplier: Spectrum™ 132480
Description
- Material: Regenerated Cellulose
- Hydrophilic
- Symmetric porosity
- Carries no fixed charge
- Does not adsorb most solutes
- pH limits: 2 to 12
- Packaged dry with glycerol (must be rinsed out with water)
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation
Specifications
RC Dialysis Disc | |
Spectra/Por 2 | |
47 mm | |
50 Pack |
12 to 14 kDa | |
Regenerated Cellulose | |
1.85 in. | |
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation |