missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

American Scientific Spectrum Discharge Tubes

Glass tubes about 26cm long, used to show spectral lines of pure substances.

Supplier:  American Scientific 337602

 Afficher plus de versions de ce produit

Numéro de catalogue. S01682B


Découvrez plus d'offres spéciales

Avertissement:

Tubes should be powered for 30 seconds and not more than two minutes (to avoid burning of bulbs).


Description

Description

  • Central capillary portion is approximately 9-10cm long with electrodes sealed at both ends and connected to lead wires protected by metal caps
  • Loops welded to caps for electrical connection
  • Needs an induction coil 5cm spark, or a suitable transformer
  • Length of capillary portion can be varied as required
  • For intermittent use, cannot be used as lamps
  • Options include: Air, Argon, Carbon Dioxide, Helium gas, Hydrogen gas, Iodine, Krypton gas, Mercury gas, Neon gas, Nitrogen gas, Oxygen, Water vapor, Xenon, Bromine, and Deuterium

Spécifications

Spécifications

Argon Spectrum Discharge Tube
Used to show spectral lines of pure substances
Electrodes sealed at both ends
10.25 in.
3.5 to 4 in.
Violet
Induction coil 5cm spark/a transformer
Glass
26 cm
8.9 to 10.16 cm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Offres spéciales

Offres spéciales