Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
Supplier:  Spectrum™ 132723
 View more versions of this product
Catalog No. 08-671-20
Explore available promotions
Description
Description
  • Flat-Sheets deal for use with dialysis equipment or cutting to size
  • Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
  • Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Specifications
Spectra/Por 3 Dialysis Membrane Flat Sheets
Spectra/Por 3
108 mm
108 mm
Clear
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
3.5 kDa
4.2 in.
4.2 in.
4.2 x 4.2 in. (108 x 108 mm)
39°F to 250°F
25 Sheets