Spectrum™ Spectra/Por™ 4 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting.
Supplier:  Spectrum™ S432706
 View more versions of this product
Catalog No. 08-667E
Explore available promotions
Description
Description
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus.
Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
Spectra/Por 4 Dialysis Tubing
Regenerated Cellulose
1.77 in.
30 m
39.2°F to 249.8°F
Each
12 to 14 kDa
45 mm
100 ft.
Packaged with glycerin
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Videos
SDS
Documents
Documents
Accessories