missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spectrum™ Standard RC Ready-to-use Spectra/Por™ Dialysis Sacks

For large sample volumes

Supplier:  Spectrum™ 132726

 View more versions of this product

Catalog No. 08-801-06


Explore available promotions

Description

Description

  • Material: Regenerated Cellulose
  • Length: 60cm (23.62 in.)
  • Sealed at one end, open at the other
  • Funnel attached to top for easy sample filling
  • Supplied in sodium azide 0.05% (must be rinsed prior to use)

Specifications

Specifications

Dialysis Sacks
Spectra/Por 3
34 mm
23.6 in.
54 mm
0.05% sodium azide preservant
For large sample volumes, funnel attached to top for easy sample filling
3.5 kDa
Regenerated Cellulose
1.33 in.
600 mm
50 to 400 mL
10 Pack
Videos
SDS
Documents

Documents

Promotions

Promotions