Honeywell™ Sperian™ Qualitative Bitrex Fit-Test Kit
Sperian offers various Qualitative (QLFT) fit test kits for all your testing needs. Kit verifies assigned protection factors of ten.
Supplier:  Honeywell™ 193170
Catalog No. 19-005185
Explore available promotions
Includes: Tyvek™ test hood, sensitivity nebulizer, fit-test nebulizer, 2 oz. (60mL) vial of threshold screening solution, 2 oz. (60mL) vial of fit-test solution and two sets of replacement nebulizer inserts.
Description
Description
Fit-test kit offers an easy and economical method of fit-testing disposable and reusable NIOSH-approved respirators, including 42CFR84 respirators with N, R, or P filters. Bitrex, an extremely bitter solution, is easy to detect by test subjects. 2 oz. (60mL) vial of test solution lasts for 10 to 20 fit tests. Training video shows and describes qualitative fit-test procedures for Bitrex and IAA (banana oil). Covers directions for a complete fit-test, including sensitivity screening, the actual fit-test and documenting fit-test findings.
Specifications
Specifications
Easy and economical method of fit-testing disposable and reusable
Tyvek™ test hood, sensitivity nebulizer, fit-test nebulizer, 2 oz. (60ml) vial of threshold screening solution, 2 oz. (60ml) vial of fit-test solution and two sets of replacement nebulizer inserts
1 Ea.