missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Spermine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132750010

 View more versions of this product

Catalog No. AC132750010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Spermine
71-44-3
0.9370g/mL
110°C
Glass bottle
H2N(CH2)3NH(CH2)4NH(CH2)3NH2
1 g
0.937
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
NCCCNCCCCNCCCN
202.35
CHEBI:15746
97%
97%
26.0°C to 30.0°C
130.0°C (0.1 mmHg)
97%
C10H26N4
MFCD00008215
04, II, 704
15, 8870
PFNFFQXMRSDOHW-UHFFFAOYSA-N
N,N'-bis(3-aminopropyl)butane-1,4-diamine
1103
202.34

Chemical Identifiers

71-44-3
202.35
PFNFFQXMRSDOHW-UHFFFAOYSA-N
1103
N,N'-bis(3-aminopropyl)butane-1,4-diamine
C10H26N4
MFCD00008215
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
CHEBI:15746
NCCCNCCCCNCCCN
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 200-754-2

RTECSNumber : EJ7175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only