Learn More
Spermine, 97%, Thermo Scientific Chemicals
$137.32 - $137.32
Chemical Identifiers
CAS | 71-44-3 |
---|---|
Molecular Formula | C10H26N4 |
Molecular Weight (g/mol) | 202.35 |
MDL Number | MFCD00008215 |
InChI Key | PFNFFQXMRSDOHW-UHFFFAOYSA-N |
Synonym | spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl |
PubChem CID | 1103 |
ChEBI | CHEBI:15746 |
IUPAC Name | N,N'-bis(3-aminopropyl)butane-1,4-diamine |
SMILES | NCCCNCCCCNCCCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC132750010
|
Thermo Scientific Chemicals
132750010 |
1 g | Glass bottle |
Each for $137.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-44-3 | |
202.35 | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
1103 | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine |
C10H26N4 | |
MFCD00008215 | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
CHEBI:15746 | |
NCCCNCCCCNCCCN |
Specifications
71-44-3 | |
0.9370g/mL | |
110°C | |
Glass bottle | |
H2N(CH2)3NH(CH2)4NH(CH2)3NH2 | |
1 g | |
0.937 | |
spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl | |
NCCCNCCCCNCCCN | |
202.35 | |
CHEBI:15746 | |
97% |
26.0°C to 30.0°C | |
130.0°C (0.1 mmHg) | |
97% | |
C10H26N4 | |
MFCD00008215 | |
04, II, 704 | |
15, 8870 | |
PFNFFQXMRSDOHW-UHFFFAOYSA-N | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine | |
1103 | |
202.34 | |
Spermine, 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat
GHS Signal Word: Danger
EINECSNumber : 200-754-2
RTECSNumber : EJ7175000
TSCA : TSCA
RUO – Research Use Only