missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SPEX™ SamplePrep Shatterbox™ Accessory, Grinding Containter
For use with Shatterbox
$6019.69 - $31187.96
Specifications
Capacity (English) | 0.67 to 1.69 oz. |
---|---|
Capacity (Metric) | 20 to 50 mL |
Includes | Lid |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
8504
|
SPEX™ SamplePrep
SPX8504 |
Tungsten Carbide |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
8501
|
SPEX™ SamplePrep
8501 |
Hardened Steel |
Each for $6,019.69
|
|
|||||
Description
Containers
- Choice of steel, carbide or ceramic styles
- Supplied with lid
Specifications
0.67 to 1.69 oz. | |
20 to 50 mL | |
Lid |