missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec Group Sphere Flash™ Economy Automated Colony Counter

Sphere Flash™ Economy Automated Colony Counter accurately counts pour and spiral plates on any media in one second with automatic calibration and verification.

Supplier:  Neutec Group 90007000

Catalog No. 14-910-200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Counts the majority of colonies on various media as an automated alternative to manual counting. The high resolution color camera detects colonies of 1/450 inch and can process any type of media, including transparent and opaque agar without ambient light interference due to closed environment.

  • Adaptable to specific micro lab needs
  • Upgradable
  • Modern LED illumination
  • Internet connectivity
  • Computer interface: USB 3.0 Type-A socket
  • Petri dish maximum usable diameter: 90mm
  • Relative humidity: 10-75% (non-condensing)
Specifications

Specifications

Automated Colony Counter
288 x 220 x 371 mm
Maximum Power:40W
Aprox. 1/450 inch (0.07mm)
5.5 Kg
For use in a variety of microbiology laboratories such as Environmental, Food, Pharma, Biotech.
Videos
SDS
Documents

Documents

Promotions

Promotions