missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eagle™ Spill Containment Drums

Approved for use with Packing Groups I, II and III

Supplier:  Eagle™ 1610MB

 View more versions of this product

Catalog No. 19-160-518


Explore available promotions

Description

Description

Lightweight drums for safe storage, transportation, cleanup and containment of containers from 5 to 55 gallons. Made of chemical- and weather-resistant HDPE with UV inhibitors that incinerate completely. Nestable for ideal storage.

Lab Packs/Open Head Drums

  • For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials
  • Color: Blue
  • Includes white lid with metal lever-lock band closure

     

 

Order Info

Lab Packs and Overpacks cannot contain free liquids. Sufficient sorbents must be used with any leaking containers to absorb all liquids.

Certifications

UN certified to HM-181. UL 94HB Flammability Rating.

Compliance

All drums meet FDA 21CFR177.1520. Lab packs meet DOT 49CFR173.12. Overpacks meet DOT 49CFR173.25. Salvage drums meet DOT 49CFR173.3.

Specifications

Specifications

DOT 49 CFR 173.3, FDA 21 CFR 177.1520
14 gal.
HDPE
Ideal for storage, transportation, clean up and spill containment of larger containers
White lid with metal lever-lock band closure
100 kg
26.5 in.
10 lb.
Blue
52.2 L
Lab Pack poly drum; Metal Lever-lock
Bottles, cans, carboys and 5 gal. pails
220 lb.
Chemical, Weather
67.3 cm
4.5 kg
SDS
Documents

Documents

Promotions

Promotions