missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Contec™ Sporicidin™ Fresh Scent Disinfectant Wipes
Sporicidin™ wipes are EPA registered for use in hospitals, medical, dental fields, meets EPAâs criteria for use against SARS-CoV-2
Supplier: Contec™ JUM8506FCAN
Description
- Used in hospital dental and medical environments since 1978
- Kills disease adn odor-causing organisms
- Broad spectrum disinfectant- no rotation of intermediate disinfectants needed
- EPA registered in all 50 states and Puerto Rico
- OSHA bloodborne pathogen standards (29 CFR 1910.1030) compliant
- EPA toxicity rating low/very low
- Cleans, disinfects and deodorizes in one step
- Kills bacteriostatic activity up to 6 months, if not removed from surface
- Fungicidal, bactericidal, virucidal, mildewcidal and tuburculocidal
- Non-corrosive to surfaces including plastics, latex, vinyl, glass, treated wood, metal and glazed porcelain
- Continas pharmaceutical active ingredient system with no harsh chemicals
- Mildly alaline (pH 8.5)
- Non-staining, non-flammable and non-abrasive to most surfaces
- Non-hazardous formula with no special shipping or disposal restrictions
- Alcohol and bleach-free wipes safe for medical and dental equipment
- Designed for heavy duty cleaning needs
- Have two distinct cleaning surfaces
- One side abrasive texture for aggressive cleaning, one side softer texture for gentle, delicate cleaning
- Dimensions: 9.5 x 12 in. (24.1 x 30.5 cm)
- EPA Registration Number: 8383-7
Specifications
Disinfectant Wipes | |
Fresh Scent | |
One side abrasive texture, one side softer texture | |
85 Wipes/Pk., 6 Pk./Cs. |
8.5 | |
EPA registered 8383-7, OSHA bloodborne pathogen standard compliant, EPA toxicity rating low/very low | |
For non-corrosive surfaces like plastics, latex, vinyl, glass, treated wood, metal and glazed porcelain surfaces in hospitals, dental and medical environments, for heavy duty cleaning |