MilliporeSigma™ Supelco™ Stainless Steel 1/16 in. Capillary Tubing, Length: 1 m
Use to connect the column to the detector compartment in a capillary gas chromatographic setup
Supplier:  MilliporeSigma™ Supelco™ 56736
 View more versions of this product
Catalog No. 111005156
Explore available promotions
Description
Description
Specifications
Specifications
Capillary Tubing
0.030 in.
1 m
Gas Chromatography (GC)
1/16 in.
Stainless Steel