Fisherbrand™ Stainless Steel Canopy Hood
Fisherbrand™ Canopy hoods are suitable for the removal of heat or nuisance fumes.
 View more versions of this product
Catalog No. 06000638
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Canopy hoods need to be tied into the general exhaust for the lab.
  • Canopy hoods have a flat back and are designed for perimeter locations.
  • Stainless steel hoods are made of Type 304 stainless steel with #4 finish and include a duct collar.
  • Canopy includes internal baffle and 48 in. threaded hanging rods.
Specifications
Specifications
Stainless Steel
61 cm
45.7 cm
72 in.
24 in.
18 in.
Internal Baffle and 48 in. Threaded Hanging Rods
182.9 cm