Fisherbrand™ Stainless Steel Cleanroom Lab Table with Chemical Resistant Phenolic Resin Top
FisherBrand™ Class 1 Cleanroom Lab Table features an adjustable height range with 2-inch increments and a 3 minute assembly
Supplier:  Fisherbrand™ KSY2472+LP
 View more versions of this product
Catalog No. 25-100-499
Explore available promotions
Description
Description
  • Grade 304 solid stainless steel frame
  • No.4 brushed finish stainless with chemical resistant phenolic resin top
  • Easy to clean
  • Meets and exceeds SEFA
Specifications
Specifications
25 year unconditional manufacturer warranty
24 in.
30 to 36 in.
Bench Frame, legs and top
609 mm
1828 mm
ANSI / BIFMA, SEFA
Stainless Steel (304)
Cleanroom Table
762 to 914 mm
30 to 36 in. (2 in. increments)
72 in.
2 in.
Stainless
5,000 lb