Troemner™ Stainless Steel Electronic Balance Calibration Weights, Class 4 with NVLAP Certificate
Precision-crafted of stainless steel
Supplier:  Troemner™ 30391175
 View more versions of this product
Catalog No. 02-100-123
Explore available promotions
Description
Description
All electronic balance weights are manufactured with adjusting cavities
  • Weights 5kg to 30kg are flat on top with a recessed grip handle for easy handling and stacking
  • Weights 1kg to 5kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping
  • Weights below 1kg are straight cylinders
  • Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove
  • With NVLAP Certificate
Specifications
Specifications
Two-Piece Electronic Balance Calibration Weight
ASTM Class 4
50 g
50 g
Polypropylene case
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with Recessed Grip
Each