missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Stainless Steel ESD Cleanroom Lab Table
FisherBrand™ Class 100 Cleanroom Lab Table features an adjustable height range with 2-inch increments and an easy 3 minute assembly
Supplier: Fisherbrand™ KSDCR3648+LP
Description
- Grade 304 solid stainless steel frame
- No.4 brushed finish stainless
- Easy to clean
- Meets and exceeds SEFA
Specifications
25 year unconditional manufacturer warranty | |
36 in. | |
30 to 36 in. | |
Bench Frame, legs and top | |
609 mm | |
1219 mm | |
ANSI / BIFMA, SEFA | |
White |
Cleanroom Table | |
762 to 914 mm | |
30 to 36 in. (2 in. increments) | |
48 in. | |
2 in. | |
Stainless | |
5,000 lb | |
Stainless Steel (304) |