missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ Stainless Steel Frit, Porosity: 20 μm

For use with 6 mL Polypropylene SPE Tubes

Supplier:  MilliporeSigma™ Supelco™ 57246U

Catalog No. 111010738


Explore available promotions

Description

Description

Specifications

Specifications

Polypropylene SPE Tubes (6 mL)
60
Frit
Stainless Steel
20 μm
Videos
SDS
Documents

Documents

Promotions

Promotions