Fisherbrand™ Stainless Steel Hand Sink
Stainless Steel Hand Sinks are constructed of heavy duty Type 304 stainless steel and include a backsplash and outer marine edge.
Catalog No. 15050323
Explore available promotions
Description
Description
  • Stainless Steel Hand Sinks are constructed of heavy-duty Type 304 stainless steel and include a backsplash and outer marine edge.
  • A faucet is provided with water flow controlled by a low voltage solenoid with an adjustable optical sensor.
  • A deep drawn positive drain bowl assures complete drainage.
  • Unit furnished with a basket drain.
  • The rear splash is 7.5 in. (191 mm) tall. Furnished complete with wall mounting brackets.
  • 120 V 15 amp electricity is required.
  • A 12 V power adaptor is included.
  • Water inlet is 0.5 in. (13 mm) and drain outlet is 1.5 in. (38 mm).
  • A thermostatic mixing valve is required.
Specifications
Specifications
Silver
Hand Sink
32.4 cm
Stainless Steel (304)
6.75 in.