missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Stainless Steel Laboratory Tripods

Stainless steel laboratory tripods can be used in a variety of laboratory set ups

Supplier:  Eisco™ CH0647B

 View more versions of this product

Catalog No. S41706


Explore available promotions

Description

Description

  • Made of plated mild steel
  • 6.5cm diameter
  • Autoclavable and chemical resistant
  • Various heights available

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Stainless Steel
34 cm
Stand
Lab stand
2.47
22 cm
25 cm
Silver
1
Videos
SDS
Documents

Documents

Promotions

Promotions