missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Stainless Steel Laboratory Tripods

Stainless steel laboratory tripods can be used in a variety of laboratory set ups

Supplier:  Eisco™ CH0647E

 View more versions of this product

Catalog No. S41709


Explore available promotions

Description

Description

  • Made of plated mild steel
  • 6.5 cm diameter
  • Autoclavable and chemical resistant
  • Various heights available

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Nickel Plated Steel Wire
8 in.
15.24 cm
Silver
1
21 cm
15.24 cm
120 mm
Lab stand
0.18
Videos
SDS
Documents

Documents

Promotions

Promotions