missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Stainless Steel Probe and Seeker

Eisco™ Stainless Steel Probe and Seeker is made form high quality stainless steel and features an angled blunt needle.

Supplier:  Eisco™ BI0179SUP

Catalog No. S29350


Explore available promotions

Description

Description

  • Probe and seeker with blunt end needle
  • Made from quality stainless steel
  • Single piece probe is 5 in. long (127 mm)
  • Great for experiments, dissections, and demonstrations

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Needle seeker
Silver
Dissection
1
12.7 cm
1.3 cm
Autoclavable
Stainless Steel
1 seeker
5 in.
1.3 cm
0.05
Videos
SDS
Documents

Documents

Promotions

Promotions