RETSCH Stainless Steel Collecting Pan with Outlet for Test Sieves
Collects resulting samples from test sieves. These stainless steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter. Outlet allows draining of liquids during wet sieving.
$500.52 - $500.52
Specifications
Type Collecting Pan With Outlet
Material Stainless Steel
For Use With (Equipment) Test sieves, Analytical Sieve Shaker AS 200 Digital
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
08-418-44
View Documents
RETSCH
694203050
203 mm
Each for $500.52
Description
Description
  • Stainless steel
  • Outlet allows draining to occur during separation or sorting
  • Compatible with multiple test sieves, based upon diameter
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks with O-ring
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
Specifications
Collecting Pan With Outlet
Stainless Steel
Test sieves, Analytical Sieve Shaker AS 200 Digital
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'