RETSCH Stainless Steel Collecting Pan for Test Sieves
Collects resulting samples from test sieves. These Stainless Steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.
$120.82 - $120.82
Specifications
Type Collecting Pan
Material Stainless Steel
For Use With (Equipment) Test Sieve
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Outer Height (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Height (Metric) Price Quantity  
08-418-46
View Documents
RETSCH
697203050
203 mm 50.8 mm
Each for $120.82
Description
Description
  • Stainless Steel
  • Compatible with multiple test sieves, based upon diameter
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks with O-ring
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
Specifications
Collecting Pan
Stainless Steel
Test Sieve
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'