Fisherbrand™ Stainless Steel Undermount Sink
Fisherbrand™ Stainless Steel Undermount Sink is a seamless die-drawn construction and mounted underneath work surface.
Includes: Crumb Strainer
Description
- 18 gauge construction
- 304 Stainless Steel
- Interior surfaces polished to a nonporous finish
- Fully coated underside insulated for sound and reduced condensation
- Self-rimming top mount Grip-Rim Plus with stainless steel mounting channels
- Centered sink outlet
- Sink supports will be required
Specifications
Stainless Steel | |
25.5 in. | |
19.25 in. | |
10 in. | |
8.2 kg |
Crumb strainer and tail piece | |
64.77 cm | |
48.89 cm | |
25.4 cm |