missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Medegen Stainless-Steel Utility Trays

Corrosion resistant and easy to clean

Supplier:  Medegen 74122

 View more versions of this product

Catalog No. 13-361B


Explore available promotions

Description

Description

Corrosion resistant and easy to clean

Specifications

Specifications

2.4 L
5.4 cm
Stainless Steel
20 cm
2.125 in.
7.375 in.
12.25 x 7.75 x 2.25 in. (31 x 20 x 5.4 cm)
31 cm
Instrument Tray
12.25 in.
Rectangular
2.5 qt.
Videos
SDS
Documents

Documents

Promotions

Promotions