Brady™ Standard Barricade Tape Roll: Non-Adhesive Lightweight Polyethylene, CONFINED SPACE DO NOT ENTER, Black on Yellow
This durable, lightweight barricade tape is made of non-adhesive, glossy B-912 polyethylene for on-the-spot warnings of potential danger.
Supplier: Brady™ 91256
Description
- Non-adhesive Polyethylene (B-912) is a glossy plastic that's ideal for temporary outdoor or permanent indoor use
- Lightweight tape can be tied, stapled or nailed to posts, fences or metal barricades
- Visual barricading provides fast on-the-spot warning of potential danger
- B-912 polyethylene barricade tapes from Brady™ are available in variety of legends to fit your specific needs (each roll/legend is sold separately)
Specifications
Black on Yellow |