missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Welch™ Standard-Duty Vacuum Pumps

Compact, economical WOB-L™ pumps for aqueous applications

Supplier:  Welch™ 2585B50

 View more versions of this product

Catalog No. 01-257-73


Explore available promotions

Alert:

May be used with buffer solutions but are not recommended for use with acids, bases or organic vapors.

Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.


Includes: Liquid trap on inlet, vacuum gauge and adjustment knob, exhaust muffler, suction cup feet, and handles for convenient transportation

Description

Description

Compact, economical WOB-L™ pumps for aqueous applications

  • Ideal for filtration, vacuum ovens, aspiration and chamber roughing
  • Protect against accidental ingestion
  • Wetted parts resist corrosion from moisture

Order Info

For 115V 60Hz. All pumps weigh less than 25 lb. (11.4kg).

Specifications

Specifications

0.33 hp
14.5 kg
CE
2585
11.1 kg
60 Hz
30.5 cm
43.1 cm
115 V
32 lb.
Liquid trap on inlet, vacuum gauge and adjustment knob, exhaust muffler, suction cup feet and handles for convenient transportation
115 V 60 Hz
24.5 lb.
201 L/min.
12 in.
17 in.
8 in.
Videos
SDS
Documents

Documents

Promotions

Promotions