Learn More
American Biotech Supply Standard Hazardous Location (Explosion Proof) Refrigerator with Natural Refrigerants
Refrigerators are engineered for cold-storage use in environments where volatile conditions could potentially exist. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.
Specifications
Type | HazLoc Refrigerator |
---|---|
Temperature Range | 1°C to 10°C |
Temperature Sensor | Microprocessor Temperature Controller |
Temperature Stability | ±0.1°C |
Refrigerant | Natural, Hydrocarbon |
Catalog Number | Mfr. No. | Energy Usage | Capacity (English) | Capacity (Metric) | Min. Clearance | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Energy Usage | Capacity (English) | Capacity (Metric) | Min. Clearance | Price | Quantity | ||
22-651-383
|
American Biotech Supply
ABTHCERP14 |
1 to 5 HP | 14 cu. ft. | 396 L | 75 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
22-651-384
|
American Biotech Supply
ABTHCERP17 |
1 to 4 HP | 17 cu. ft. | 481 L | 70 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
22-651-385
|
American Biotech Supply
ABTHCERP20 |
1 to 4 HP | 20 cu. ft. | 566 L | 80 in. |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Standard Hazardous Location (Explosion Proof) Refrigerators with Natural Refrigerants are engineered for cold-storage use in environments where volatile conditions could potentially exist. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. These units meet NFPA and OSHA guidelines Class 1, Division II, Groups C and D for storing substances in hazardous locations.*
*Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see ABS Flammable Storage Units.
- Microprocessor Temperature Controller
- Interior shelving
- Keyed door lock
- High density urethane foam cabinet and door insulation
- Not equipped with line cord and plug; must be hardwired in conduit to 120 VAC power source
- ETL Listed
Specifications
HazLoc Refrigerator | |
Microprocessor Temperature Controller | |
Natural, Hydrocarbon | |
1 | |
5 Adjustable | |
Powder-coated Steel | |
Plastic | |
Upright | |
White | |
Cold Storage | |
Two Year Parts and Labor Warranty | |
Microprocessor Temperature Controller | |
3 A | |
115 V |
1°C to 10°C | |
±0.1°C | |
Solid Swing | |
Right | |
Scratch-resistant Painted Steel | |
Polyurethane Foam | |
cULus | |
Manual | |
Fan-forced | |
None | |
Cycle | |
Bottom-mounted | |
NEMA 5-15 P |