METTLER TOLEDO Standard MA Analytical Balance
METTLER TOLEDO Standard MA Analytical Balances are highly accurate, efficient, simple to operate and designed with robust construction for proven long lasting performance.
Supplier:  METTLER TOLEDO 30666316
 View more versions of this product
Catalog No. 01-804-258
Explore available promotions
Description
Description
The LCD hybrid touchscreen is simple to operate and will increase efficiency of your daily tasks with 9 built-in applications and statistical data analysis. Built with a metal base, PBT top housing and overload protection to defend against rough handling in any weighing environment, ensuring quality performance for years to come.
MA Analytical Balances include:
  • Internal adjustment
  • LCD hybrid touchscreen with onscreen instructions
  • 9 built-in applications
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
The MA Balance is the successor to the ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications
Specifications
USB-A, RS232
Touchscreen
Metal, PBT
4 sec.
100/240 V, 50/60 Hz
52 g
0.01 mg
MA55
Analytical Balance
No