Learn More
METTLER TOLEDO Standard MA Precision Balance
METTLER TOLEDO Standard MA Precision Balances are highly accurate, simple to operate and built with high quality materials for proven long lasting performance.
$2163.05 - $9460.00
Specifications
Connection Type | USB-A, RS232 |
---|---|
Display Type | Touchscreen |
Material | Metal, PBT |
Type | Precision Balance |
Electrical Requirements | 100/240 V, 50/60 Hz |
Catalog Number | Mfr. No. | Capacity | Readability | Model | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Readability | Model | Price | Quantity | |||
01-804-248
|
METTLER TOLEDO
30697420 |
120 g | 1 mg | MA103 |
Each for $2,327.61
|
|
|||
01-804-246
|
METTLER TOLEDO
30697423 |
220 g | 1 mg | MA203 |
Each for $2,507.12
|
|
|||
01-804-244
|
METTLER TOLEDO
30697426 |
320 g | 1 mg | MA303 |
Each for $2,685.76
|
|
|||
01-804-242
|
METTLER TOLEDO
30697429 |
520 g | 1 mg | MA503 |
Each for $3,043.92
|
|
|||
01-804-240
|
METTLER TOLEDO
30697440 |
620 g | 10 mg | MA602 |
Each for $2,163.05
|
|
|||
01-804-238
|
METTLER TOLEDO
30697443 |
1200 g | 10 mg | MA1002 |
Each for $2,523.83
|
|
|||
01-804-236
|
METTLER TOLEDO
30697446 |
2200 g | 10 mg | MA2002 |
Each for $2,764.08
|
|
|||
01-804-234
|
METTLER TOLEDO
30697449 |
3.2 kg | 10 mg | MA3002 |
Each for $2,884.64
|
|
|||
01-804-232
|
METTLER TOLEDO
30697452 |
4.2 kg | 10 mg | MA4002 |
Each for $3,004.32
|
|
|||
01-804-230
|
METTLER TOLEDO
30697455 |
6.2 kg | 10 mg | MA6002 |
Each for $3,305.27
|
|
|||
01-804-228
|
METTLER TOLEDO
30697458 |
5.2 kg | 0.1 g | MA5001 |
Each for $2,366.32
|
|
|||
01-804-226
|
METTLER TOLEDO
30697475 |
12.2 kg | 0.1 g | MA12001L |
Each for $7,981.61
|
|
|||
01-804-224
|
METTLER TOLEDO
30697478 |
16.2 kg | 0.1 g | MA16001L |
Each for $8,572.95
|
|
|||
01-804-222
|
METTLER TOLEDO
30697481 |
32.2 kg | 0.1 g | MA32001L |
Each for $9,460.00
|
|
|||
01-804-220
|
METTLER TOLEDO
30697484 |
32.2 kg | 1 g | MA32000L |
Each for $7,568.00
|
|
|||
Description
The precise weigh cell and internal adjustment provide accurate results. Intuitive onscreen guidance within the LCD hybrid touchscreen allows you to easily navigate through 10 built in applications and manage weighing data. The metal base and overload protection defend the balance against rough handling in any weighing environment.
MA Precision Balances include:
- Internal adjustment
- LCD hybrid touchscreen
- 10 built-in applications
- Statistical data analysis
- Front level indicator
- Metal base, PBT top housing
- Overload protection
- 2 interfaces: USB-A, RS232
- Bluetooth option
The MA Balance is the successor to the ME-T Balance.See how it ranks among our balance lines:
- XPR Excellence Precision Balances
- MX Advanced Precision Balances
- MR Advanced Precision Balances
- MA Standard Precision Balances
- ME Standard Precision Balances
- LA Standard Precision Balances
Specifications
USB-A, RS232 | |
Metal, PBT | |
100/240 V, 50/60 Hz |
Touchscreen | |
Precision Balance | |
No |