Synthware™ Standard Vacuum Trap
Deisgned to allow easy removal of condensate. Body length is measured below the joint.
Supplier:  Synthware™ V103029
 View more versions of this product
Catalog No. 31501824
Explore available promotions
Description
Description
Specifications
Specifications
32 mm
200 mm
Borosilicate Glass
29/42
Organic Synthesis
Vacuum Trap
1 Ea.