Synthware™ Standard Vacuum Trap
Deisgned to allow easy removal of condensate. Body length is measured below the joint.
Supplier:  Synthware™ V105050
 View more versions of this product
Catalog No. 31501828
Explore available promotions
Description
Description
Specifications
Specifications
55 mm
275 mm
Borosilicate Glass
50/50
Organic Synthesis
Vacuum Trap
1 Ea.