missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Stannic Oxide, Powder, 99.9%, Spectrum™ Chemical
$186.29 - $19145.94
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18614017
|
Spectrum Chemical Mfg Cor
S153025GM |
25 g | Poly Bottle |
Each for $186.29
|
|
||||
18614019
|
Spectrum Chemical Mfg Cor
S1530500GM |
500 g | Poly Bottle |
Each for $986.15
|
|
||||
18614020
|
Spectrum Chemical Mfg Cor
S153025KG |
2.5 kg | Poly Pail |
Each for $3,166.06
|
|
||||
18614016
|
Spectrum Chemical Mfg Cor
S153012KG |
12 kg | Poly Pail |
Each for $6,999.79
|
|
||||
18614018
|
Spectrum Chemical Mfg Cor
S153045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
18282-10-5 |
Specifications
18282-10-5 | |
25 g | |
Reagent |
1 | |
99.9% | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.