missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Iodometric Titrations
Supplier: Ricca Chemical Company 80001
Specifications
| Starch Indicator | |
| 0.5% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 5 to 7 | |
| Liquid |
| 9005-84-9,69-72-7,7732-18-5 | |
| 0.5% W/V | |
| Natural Poly Bottle | |
| White | |
| 4 L |