missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch, Potato, Powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 419695000

 View more versions of this product

Catalog No. AC419695000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White
6 to 8 (50% aq. suspension)
9005-25-8
C12H22O11
14, 8798
Solubility in water: insoluble.
GUBGYTABKSRVRQ-UHFFFAOYNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
439341
Extra Pure
20.5% max. (2 g, 130°C, 90 min)
Starch
Powder
0.5% max. (900°C)
MFCD00082026,MFCD00132834
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
500 g
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
342.30
CHEBI:18167
Authentic
Plastic bottle

Chemical Identifiers

9005-25-8
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
439341
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C12H22O11
MFCD00082026,MFCD00132834
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
CHEBI:18167
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Safety and Handling

Safety and Handling

EINECSNumber : 232-679-6

RTECSNumber : GM5090000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only