missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starch, Soluble Potato, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor 400605

 View more versions of this product

Catalog No. 02004319

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

    For Iodometry
Specifications

Specifications

Starch
9005-84-9
C12H22O11
1L = 1.50kg
GUBGYTABKSRVRQ-ASMJPISFSA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
439341
BAKER ANALYZED™ ACS Reagent
Soluble Potato
Poly Bottle
2.5 kg
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
342.297
CHEBI:18167
Powder

Chemical Identifiers

9005-84-9
342.297
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
CHEBI:18167
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
C12H22O11
GUBGYTABKSRVRQ-ASMJPISFSA-N
439341
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
Videos
SDS
Documents

Documents

Promotions

Promotions