missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Starch, Soluble (Powder/Certified ACS), Fisher Chemical™
For iodometry
$343.83 - $855.92
Chemical Identifiers
CAS | 9005-84-9 |
---|---|
Molecular Formula | C12H22O11 |
Molecular Weight (g/mol) | 342.297 |
MDL Number | MFCD00082026 |
InChI Key | GUBGYTABKSRVRQ-ASMJPISFSA-N |
Synonym | alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose |
PubChem CID | 439341 |
ChEBI | CHEBI:18167 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
S516-100
|
Thermo Fisher Scientific
S516100 |
100 g | Glass Bottle |
Each for $343.83
Case of 6 Each for $2,062.98
|
|
||||
S516-500
|
Thermo Fisher Scientific
S516500 |
500 g | Glass Bottle |
Each for $855.92
Case of 6 Each for $5,135.52
|
|
||||
Chemical Identifiers
9005-84-9 | |
342.297 | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
439341 | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
C12H22O11 | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
CHEBI:18167 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Specifications
White | |
Powder/Solid | |
256°C | |
MFCD00082026 | |
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
GUBGYTABKSRVRQ-ASMJPISFSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
439341 | |
Certified ACS | |
0.4% max. | |
Pass Test |
5.0 to 7.0 | |
9005-84-9 | |
C12H22O11 | |
1.0384g/cm³ | |
100 g | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:18167 | |
Pass Test | |
Glass Bottle | |
Starch, Soluble |