missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Starch Solution (Stabilized), Fisher Chemical™
For iodometry
Supplier: Fisher Chemical SS4084
Description
For iodometry
Specifications
Colorless | |
Stabilized | |
9005-25-8,64-19-7,7732-18-5 | |
0°C | |
MFCD00082026 | |
4 L | |
Pass Test |
Starch Solution | |
Liquid | |
100°C | |
(C6H10O5)n | |
1.0g/cm³ | |
Poly Bottle | |
14mmHg |