missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Starplex™ Scientific LeakBuster™ Specimen Containers: Sterile

Ideal for use in harsh transport conditions or in pneumatic tube systems

Supplier:  Starplex™ Scientific NCS6021O

 View more versions of this product

Catalog No. 09-735-201


Explore available promotions

Description

Description

Ideal for use in harsh transport conditions or in pneumatic tube systems

A premium line of containers designed for leakproof transportation.

  • Streamlined straight-sided vials are stable, stackable, and save space in storage
  • Caps are designed with a full-circle thread that prevents the cap from cross threading and leaking
  • QuickTurn™ caps are provided on the wide-mouth containers; lids are secured with only a quarter of a turn
  • Sterile containers have an orange cap and tamper tab patient identification label that meets current JCAHO standards
  • LeakBuster containers have passed DOT/ICAO/IATA 95kPa Pressure Differential and Thermal Shock requirements

Order Info

Individual sterile containers have large identification labels and tamper-evident tabs.

Specifications

Specifications

Polypropylene
2 oz.
Orange
Screw Cap with O-Ring
Standard opening
Yes
Sterile
60 mL
48 mm
White
4 oz. (119 mL) specimen container
500/Cs.
Videos
SDS
Documents

Documents

Promotions

Promotions