missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Stationary Vehicle Placards, FLAMMABLE GAS 2
Indicate delivery vehicles carrying dangerous or hazardous materials. Brady™ Stationary Vehicle Placards are available in a variety of materials.
$4.32 - $4.32
Specifications
| Adhesive Type | Non-adhesive |
|---|---|
| Color | White, Red |
| Product Type | Vehicle Placard |
| Display Text | FLAMMABLE GAS 2 |
| For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
| Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
|
19-111-346
|
Brady™
63442 |
Polycoated Tagstock (B-101) |
Each for $4.32
|
|
|||||
Description
- Available in two different materials Polycoated Tagstock and Premium Fiberglass
Specifications
| Non-adhesive | |
| Vehicle Placard | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond |
| White, Red | |
| FLAMMABLE GAS 2 | |
| 1 Ea. | |
| 10.75 x 10.75 in. |