Watson Gloves Stealth Black Widow Cut A6 Gloves
Stealth Black Widow Cut A6 Gloves' HPPE/steel/glass/nylon/spandex shell is cut resistant, offering excellent ANSI A6 protection.
Supplier:  Watson Gloves 384X
 View more versions of this product
Catalog No. 17-099-948
Explore available promotions
Description
Description
  • HPPE/steel/glass/nylon/spandex shell
  • Lightweight polyurethane coated palm
  • Snug-fitting knit wrist
Specifications
Specifications
ANSI Cut, ANSI Puncture
Palm
Nylon Spandex
Gray, Black
Ambidextrous
HPPE, Nylon, Spandex, Polyurethane
X-Large
Light Industrial, Warehouse, Sheet Metal
Gray
Unlined
384
Stealth
9.5 in.
EN388
Seamless Knit
Polyurethane
ANSI A6
Yes
Cut-resistant Gloves
EN388:4X42F
Men's
Continuous Knit
Unlined
ANSI/ISEA 105-16 Level 3
13
23 cm