missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Stem-Cell Factor/c-Kit Inhibitor
Description
Inhibits in vitro phosphorylation of c-Kit. Also inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Molecular Weight (g/mol) | 355.50 |
| Quantity | 10mg |
| Packaging | Plastic bottle |
| Sensitivity | Light sensitive |
| Form | Solid |
RUO – Research Use Only