Gibco™ StemPro™ BM Mesenchymal Stem Cells DFS Item
StemPro™ BM Mesenchymal Stem Cells
Supplier:  Gibco™ A15652
Catalog No. A15652
Explore available promotions
Description
Description
StemPro™ BM Mesenchymal Stem Cells are cryopreserved human bone marrow mesenchymal stem cells (BM MSCs). These Off-the-Shelf cells are manufactured to meet Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemia tolerant (it)) proprietary clinical-scale manufacturing process using Bovine Growth Serum (BGS)-supplemented media and cryopreserved in an animal origin free cryopreservation medium. The cells are supplied at 1x106 and 5x106 viable cells/vial. A Certificate of Analysis is available for each lot which documents the quality control specifications, test results and donor information.
StemPro™ BM Mesenchymal Stem Cells allow you to:
  • Minimize assay variability with itMSCs manufactured in compliance with GMP manufacturing standards
  • Improve functionality studies with immature, high potency MSCs
  • No observed tumorigenicity and toxicity in GLP-compliant animal studies
  • Accommodate experiments requiring one large lot of MSCs
  • Minimize your chance of assay failure when using tested cells, reagents and protocols
    Immature, High Potency, Low Oxygen-Cultured MSCs
    StemPro™ BM Mesenchymal Stem Cells are derived from human bone marrow aspirates from qualified, traceable donors. The cells are isolated, cultured and expanded in compliance with GMP manufacturing standards in a California-licensed facility using a proprietary clinical-scale Reduced Oxygen Tension manufacturing process. Cells manufactured in reduced oxygen tension environments result in higher cell yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.
    Proven Non-toxic Profiles
    Cells have demonstrated to be non-toxic and non-tumorigenic in GLP animal studies, which were performed according to FDA guidelines.
    A large lot can be made available for those requiring assay standardization. Protocols using tested reagents will be provided to help minimize your chance of assay failure when using reagents and cells that have not been evaluated to work together. View our tested media and reagents.
    High Post-Cryopreservation Viability
    StemPro™ Mesenchymal Stem Cells are supplied in CryoStor™ cryopreservation medium at 1x106 and 5x106 viable cells/vial. This medium is animal origin free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling and shipping to help minimize risk of adverse effects to the quality of the cells.
    Made in USA by Stemedica for Thermo Fisher Scientific.
  • Specifications
    Specifications
    Passage 4
    Adult
    Adherent Cell Culture
    Human
    Normal
    1 x 106 Cells
    1 x 106 cells
    1 vial. Remove from foil pouch and store in liquid nitrogen.
    Stem Cell (Mesenchymal)
    Cryopreserved
    Bone Marrow
    Serum-Supplemented or Serum-Free Media
    StemPro™
    Videos
    SDS
    Documents
    Documents
    Promotions
    Promotions
    For Research Use Only. Not for use in diagnostic procedures.