Mopec™ Sterile Scalpel Blades
High-quality, stainless steel, sterile scalpel blades used for anatomic pathology or autopsy. Available in a variety of sizes.
Supplier: Mopec™ AJ211
Description
- Individually wrapped
Specifications
Stainless Steel | |
# 22 | |
Sterile | |
No |
Scalpel | |
Single-use | |
100 Blades/Pk. |