Greiner Bio-One Sterile Swab Tubes
Sterilized by irradiation. Greiner Bio-One Sterile Swab Tubes is suitable for collection and transport of non-human bacteriological, serological and cytological samples.
Supplier: Greiner Bio-One 420180
Description
- Sterilized by irradiation
- Material: Polystyrene
- For hygienic controls in food industry and environmental sampling
Specifications
Swab Tube | |
Collection and transport of non-human bacteriological, serological and cytological samples | |
1000/Cs. |
Sterile | |
110 mm | |
16 mm |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated