SP Bel-Art™ Sterileware Jumbo Sampling Pipettes
Ideal for sampling from narrow- or wide-mouth containers up to 208-liter (55 Gallon) drums
$1695.90 - $2289.79
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
01184184
|
SP Bel-Art™
F378720900 |
180 mL |
Pack of 50 Each for $2,289.79
|
|
|||||
01184185
|
SP Bel-Art™
F378721100 |
225 mL |
Pack of 37 Each for $1,695.90
|
|
|||||
Description
- Reaches into container while keeping hand clear of contents
- Diameter allows for use through drum bunghole or other small openings
- Color-coded end helps prevent handling errors
- Low-viscosity liquids enter easily through the 3 mm (1â8 in.) tip hole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) pipettes heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
2.1 cm |